1-Pentanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Pentyl alcohol, 1-Hydroxy pentane, n-Amyl alcohol, Amyl alcohol, Butyl carbinol
76929-1L 76929-250ML
SIAL76929-1LEA 164 EUR
SIAL76929-1L SIAL76929-250ML
1-Pentanol, Sigma-Aldrich®
1-Pentanol
Formula: H₃C(CH₂)₃CH₂OH
Boiling Pt: 137.9 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00002977
CAS Number: 71-41-0
EINECS: 200-752-1
UN: 1105
ADR: 3,III
Merck Index: 13,07195
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92dac1192eb8e068fc1804fdcf488129a2385918&vid=d4c7f2c5672a97c6dfaddc50807d768068096252&prid=web&clid=SAPDAM