(±)-3-Heptanol 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-Heptyl alcohol, 3-Heptanol, Heptan-3-ol, Butylethylcarbinol, (±)-Ethyl butyl carbinol, (±)-Butyl ethyl carbinol, (±)-3-Hydroxy heptane
149091000 149095000 149090250
ACRO149091000EA 131 EUR
ACRO149091000 ACRO149095000 ACRO149090250
(±)-3-Heptanol 98%
(±)-3-Heptanol
Formula: C₇H₁₆O
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –70 °C
Density: 0.818 g/cm³
Flash Pt: 55 °C
Storage Temperature: Ambient
MDL Number: MFCD00004586
CAS Number: 589-82-2
EINECS: 209-661-1
UN: 1987
ADR: 3,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4220 (20°C, 589 nm)
GC ≥97.5 %