Glycerine 99.5+% for spectroscopy
Supplier: Thermo Fisher Scientific
Synonyms: 1,2,3-Propanetriol, Propane-1,2,3-triol, Glycerol
184695000 184690025
ACRO184695000EA 99.5 EUR
ACRO184695000 ACRO184690025
Glycerine 99.5+% for spectroscopy
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1.261 g/cm³
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.5 % (Coulometric)
GC ≥99.50 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA