Glycerine ≥99.5% ACS
Supplier: Thermo Fisher Scientific
|
Warning
|
36646.AP
36646.K2
36646.M1
36646.APEA
67.7
EUR
36646.AP
36646.K2
36646.M1
Glycerine ≥99.5% ACS
Glycerine
Purity: 99.5+%
CAS No.: 56-81-5
Molecular Formula: C3H8O3
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colourless
Odour: Odourless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 182 °C (20 mmHg) Melting Pt: 18 °C Density: 1.26 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |