1,9-Nonanedithiol 97
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 1,9-Dimercaptononane
L07172.03 L07172.06
L07172.03EA 56.5 EUR
L07172.03 L07172.06
1,9-Nonanedithiol 97
1,9-Nonanedithiol
Purity: 97% CAS No.: 3489-28-9 Molecular Formula: HS(CH2)9SH Formula Weight: 192.39 Flash Point: 100°(212°F) Refractive Index: 1.4999 Boiling Point: 351-352° Beilstein Reference: 1737568 MDL No.: MFCD00004914
Formula: C₉H₂₀S₂
MW: 192.39 g/mol
Boiling Pt: 351…352 °C
Melting Pt: –17.5 °C
Density: 0.953
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004914
CAS Number: 3489-28-9
EINECS: 222-482-3