Sodium dodecyl sulphate (SDS)
Supplier: Roth Carl
|
Danger
|
Synonyms:
Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
2326.2
2326.1
2326.3
ROTH2326.2EA
0
EUR
ROTH2326.2
ROTH2326.1
ROTH2326.3
Sodium dodecyl sulphate (SDS)
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 204…207 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,II Merck Index: 14,08636 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...