Sodium dodecyl sulphate (SDS), Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

71729-1KG 71729-100G 71729-500G
SIAL71729-1KGEA 678 EUR
SIAL71729-1KG SIAL71729-100G SIAL71729-500G
Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 II
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR