n-Hexane, anhydrous
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Hexyl hydride, Hexanen, Hexane
43263.K2
43263.AK
43263.K2EA
170
EUR
43263.K2
43263.AK
n-Hexane, anhydrous
n-Hexane
CAS No.: 110-54-3
Molecular Formula: C6H14
Formula Weight: 86.18
Flash Point: -23°(-9°F)
Physical Form: Liquid
Density: 0.659
Refractive Index: 1.3750
Appearance: Colourless
Odour: Weak, characteristic
Melting Point: -95°
Boiling Point: 69°
Beilstein Reference: 1730733
Merck Reference: 14,4694
UN No.: 1208
MDL No.: MFCD00009520
Solvent in Sol-Gel precursor applications
Formula:
H₃C(CH₂)₄CH₃ MW: 86.18 g/mol Boiling Pt: 69 °C Melting Pt: –95 °C Density: 0.659 Flash Pt: –23 °C (–9 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009520 CAS Number: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3 II Merck Index: 14,4694 |