n-Octacosane 99+%
Supplier: Thermo Fisher Scientific
Synonyms:
N-octacosane
129270050
129270250
129271000
129275000
ACRO129270050EA
17.6
EUR
ACRO129270050
ACRO129270250
ACRO129271000
ACRO129275000
n-Octacosane 99+%
n-Octacosane
Formula:
H₃C(CH₂)₂₆CH₃ Boiling Pt: 440 °C (760 mmHg) Melting Pt: 61…64 °C Density: 0.8067 g/cm³ Flash Pt: 227 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009355 CAS Number: 630-02-4 EINECS: 211-125-7 |
Specification Test Results
Appearance | White Glistening powder or flakes |
Infrared spectrum | Conforms |
Melting point | 61 to 64 °C |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...