Propiolamide 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
H52319.MD
H52319.03
H52319.06
H52319.MDEA
79
EUR
H52319.MD
H52319.03
H52319.06
Propiolamide 96%
Propiolamide
Purity: 96%
CAS No.: 7341-96-0
Molecular Formula: C3H3NO
Formula Weight: 69.06
MDL No.: MFCD04035573
Formula:
C₃H₃NO Storage Temperature: Ambient |
MDL Number:
MFCD04035573 CAS Number: 7341-96-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...