Cobalt(II) thiocyanate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Cobalt dirhodanide, Cobaltous rhodanide, Cobalt(II) thiocyanate, Cobalt dithiocyanate, Cobaltous thiocyanate
M-Clarity™ quality level = MQ100
497274-25G 497274-5G
SIAL497274-25GEA 325 EUR
SIAL497274-25G SIAL497274-5G
Cobalt(II) thiocyanate, Sigma-Aldrich®
Cobalt (II) thiocyanate
Formula: C₂CoN₂S₂
MW: 175.1 g/mol
Density: 1.955 g/cm³ (25 °C)
MDL Number: MFCD00010937
CAS Number: 3017-60-5
EINECS: 221-156-8