Propagyl bromide 80% ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Bromopropyne
M-Clarity™ quality level = MQ200
81831-250ML 81831-50ML
SIAL81831-250MLEA 385 EUR
SIAL81831-250ML SIAL81831-50ML
Propagyl bromide 80% ∼9.2 mol/l in toluene ≥80%, Sigma-Aldrich®
Propagyl bromide
Formula: C₃H₃Br
MW: 118.96 g/mol
Boiling Pt: 90
Melting Pt: -43
MDL Number: MFCD00000241
CAS Number: 106-96-7
EINECS: 203-447-1
UN: 2924
ADR: 3 II