n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
Supplier: Thermo Fisher Scientific
Danger
326640010 326640025
ACRO326640010EA 89.8 EUR
ACRO326640010 ACRO326640025
n-Pentane 99+%, ECD tested for pesticide analysis for residue analysis
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 35…36 °C (1013 hPa)
Melting Pt: –130 °C
Density: 0.626 g/cm³
Flash Pt: –49 °C
Storage Temperature: Ambient
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.005 % (Coulometric)
GC ≥98.5 %
Color scale ≤5 APHA
Residue after evaporation ≤0.0005 %
Acidity (CH3COOH) ≤0.003 %