Disposable earplugs, E-A-RSoft™
Supplier: 3M
ES01001 ES01005 PD01010
111-1630EA 74.8 EUR
111-1630 111-1797 111-2422
Disposable earplugs, E-A-RSoft™
Earplugs Disposable Earplugs
E-A-RSoft™ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexibility and a good seal with optimum comfort. Once inserted in the ear, the foam plugs expand to provide a snug and secure custom fit. These roll down earplugs are available with and without cord and be used with the One Touch™ dispenser.
  • Soft hypoallergenic foam material for lower pressure inside the ear
  • Smooth, dirt resistant surface for better hygiene, durability and comfort
  • Polyester cord helps prevent hearing loss
  • One size that fits most ear canals
SNR: 37 dB
Certifications: EN 352-2