2,2,4-Trimethylpentane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
M-Clarity™ quality level = MQ100
360066-1L 360066-100ML
SIAL360066-1LEA 144 EUR
SIAL360066-1L SIAL360066-100ML
2,2,4-Trimethylpentane, Sigma-Aldrich®
2,2,4-Trimethylpentane
Formula: (CH₃)₂CHCH₂C(CH₃)₃
MW: 114.23 g/mol
Boiling Pt: 99.2
Melting Pt: –107 °C
Density: 0.692 g/cm³ (20 °C)
Flash Pt: –12 °C
Storage Temperature: Ambient
MDL Number: MFCD00008943
CAS Number: 540-84-1
EINECS: 208-759-1
UN: 1262
ADR: 3 II
Merck Index: 13,05212
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57496edad4db111238ada6124c3cea206ebf33e0&vid=bb9d5eb96909b6bebc6a8709db78c4ea0ef52430&prid=web&clid=SAPDAM