Reusable earplugs, E-A-R™ Tracer™
Supplier: 3M
TR01000 TR01001 TR01020
111-0726EA 236 EUR
111-0726 111-1128 111-1129
Reusable earplugs, E-A-R™ Tracer™
Earplugs Reusable Earplugs
The unique, one size E-A-R™ Tracer™ earplugs fit most ear canals comfortably and provide high levels of attenuation with the added benefit of incorporating a metal detectable cord which makes it ideal for the food manufacturing industry. The pre-moulded, triple-flanged plugs are made from flexible materials that are pre-formed to fit the ear.
  • Washable, reusable, easy to clean
  • Longer stem helps make insertion easier
  • Made from soft, durable silicone rubber with cord
SNR: 32 dB (Tracer™)/20 dB (Tracer™ 20).
Certifications: EN 352-2
Caution: * One size fits all.