Squalane, Sigma-Aldrich®
Supplier: Merck
85630-1L 85630-250ML
SIAL85630-1LEA 304 EUR
SIAL85630-1L SIAL85630-250ML
Squalane, Sigma-Aldrich®
Squalane
Formula: C₃₀H₆₂
Boiling Pt: 470 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.758 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008953
CAS Number: 111-01-3
EINECS: 203-825-6
Merck Index: 12,08923