2-Methylpentane, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,1-Dimethylbutane, Isohexane
68312-100ML 68312-500ML
SIAL68312-100MLEA 71.6 EUR
SIAL68312-100ML SIAL68312-500ML
2-Methylpentane, Sigma-Aldrich®
2-Methylpentane
Formula: C₆H₁₄
Boiling Pt: 62 °C (1013 hPa)
Melting Pt: –154 °C
Density: 0.653 g/cm³ (20 °C)
Flash Pt: –33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009406
CAS Number: 107-83-5
EINECS: 203-523-4
UN: 1208
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce61b1646b637970765dc4b16a997650aeccfd1&vid=71606860b34952f73fd29b7df735dd0916b1e93c&prid=web&clid=SAPDAM