Chromatographic columns with indentations and PTFE stopcock
Supplier: Lenz Laborglas GmbH & CO.KG
05440704 05440702 05440706 05435702 05435704 05435706 05435708 05440701 05435701 05440708
552-5411EA 69.5 EUR
552-5411 552-5410 552-5431 LENZ05435702 201-0591 LENZ05435706 LENZ05435708 LENZ05440701 LENZ05435701 LENZ05440708
Chromatographic columns with indentations and PTFE stopcock
Chromatography Columns
Basic chromatographic DURAN® glass columns with indentations above the stopcock for placing a cotton plug, with beaded rim or NS socket, with PTFE stopcock and retaining device.