Tetrachloroethylene
Supplier: LGC Standards PROMOCHEM
PROOHDF12EA 390 EUR
PROOHDF12
Tetrachloroethylene
Tetrachloroethylene
Formula: C₂Cl₄
MW: 165,83 g/mol
Boiling Pt: 121,1 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1,63 g/cm³ (20 °C)
MDL Number: MFCD00000834
CAS Number: 127-18-4
UN: 1897
ADR: 6.1,III
Merck Index: 12,09332
https://ru.vwr.com/stibo/web/std.lang.all/12/47/741247.jpg