trans-1,4-Dichloro-2-butene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

324515-25G
SIAL324515-25GEA 221 EUR
SIAL324515-25G
trans-1,4-Dichloro-2-butene, Sigma-Aldrich®
trans-1,4-Dichloro-2-butene
Formula: C₄H₆Cl₂
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: 1 °C
Density: 1.1873 g/cm³ (20 °C)
Storage Temperature: Refrigerator
CAS Number: 110-57-6
EINECS: 203-779-7
UN: 3390
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR