1-Dodecene, Sigma-Aldrich®
Supplier: Merck
44148-10ML 44148-50ML
SIAL44148-10MLEA 107 EUR
SIAL44148-10ML SIAL44148-50ML
1-Dodecene, Sigma-Aldrich®
1-Dodecene
Formula: C₁₂H₂₄
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.7582 g/cm³ (20 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4