1-Hexadecene ≥94%

Supplier: Thermo Fisher Scientific

L02637.AK L02637.AU
L02637.AKEA 47.1 EUR
L02637.AK L02637.AU
1-Hexadecene ≥94%
1-Hexadecene

Purity: 94% CAS No.: 629-73-2 Molecular Formula: CH3(CH2)13CH=CH2 Formula Weight: 224.43 Flash Point: 132°(269°F) Refractive Index: 1.4420 Melting Point: 4° Boiling Point: 273-274° Beilstein Reference: 1753401 MDL No.: MFCD00008991

Formula: C₁₆H₃₂
Boiling Pt: 273…274 °C
Melting Pt: 4 °C
Density: 0.781
Flash Pt: 132 °C (269 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008991
CAS Number: 629-73-2
EINECS: 211-105-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR