Cover glasses
Supplier: HIRSCHMANN
8000101 8000102 8000103 8000104 8000105 8000108 8000109 8000110 8000111 8000112 8000115 8000116 8000117 8000118 8000119 8000120
HIRS8000101EA 28.45 EUR
HIRS8000101 HIRS8000102 HIRS8000103 HIRS8000104 HIRS8000105 HIRS8000108 HIRS8000109 HIRS8000110 HIRS8000111 HIRS8000112 HIRS8000115 HIRS8000116 HIRS8000117 HIRS8000118 HIRS8000119 HIRS8000120
Cover glasses
Cover Glasses
Square or rectagular cover glasses for microscopy use.
Certifications: DIN 58 884 and IVD 98/79
Packaging: Labelled with lot number.