Disposable earplugs, E-A-R™ Classic™
Supplier: 3M
AM01000 PP01002 FP01000 CC01000
111-1060EA 106 EUR
111-1060 111-0934 111-0837 111-1067
Disposable earplugs, E-A-R™ Classic™
Earplugs Disposable Earplugs
These roll-down earplugs are made from a soft, energy-absorbing polymer foam which provides excellent hearing protection and all-day comfort.
  • Firm material with slow recovery time helps to achieve a good fit
  • Sweat and humidity resistant which improves comfort
  • Very economical as the earplugs can be cleaned up to three times
Suitable for use where noise is a problem either at work or leisure activities.
SNR: 28 dB
Certifications: EN 352-2.
Caution: * One size fits all.