1-Heptyne 99%

Supplier: Thermo Fisher Scientific
Danger

223460250 223461000
ACRO223460250EA 68.6 EUR
ACRO223460250 ACRO223461000
1-Heptyne 99%
1-Heptyne
Formula: C₇H₁₂
MW: 96.17 g/mol
Boiling Pt: 100
Melting Pt: –81 °C
Density: 0.73 g/cm³
Flash Pt: –002 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009529
CAS Number: 628-71-7
EINECS: 211-051-5
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow-green Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4090 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR