1-Tridecyne ≥97%

Supplier: Thermo Fisher Scientific
Warning

L02481.03 L02481.06
L02481.03EA 56 EUR
L02481.03 L02481.06
1-Tridecyne ≥97%
1-Tridecyne

Purity: 97% CAS No.: 26186-02-7 Molecular Formula: C13H24 Formula Weight: 180.34 Flash Point: 75°(167°F) Refractive Index: 1.4380 Boiling Point: 116-118°/10mm Beilstein Reference: 1744658 MDL No.: MFCD00015074

Formula: C₁₃H₂₄
Boiling Pt: 116…118 °C (10 mmHg)
Melting Pt: 2.5 °C
Density: 0.780
Flash Pt: 75 °C (167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015074
CAS Number: 26186-02-7
EINECS: 247-511-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR