4-Octyne ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
43157.14
43157.06
43157.14EA
112
EUR
43157.14
43157.06
4-Octyne ≥99%
4-Octyne
Purity: 99%
CAS No.: 1942-45-6
Molecular Formula: CH3CH2CH2C≡CCH2CH2CH3
Formula Weight: 110.20
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 0.7510
Refractive Index: 1.4236
Odour: Petroleum-like
Boiling Point: 131-132°
UN No.: 3295
MDL No.: MFCD00009471
Formula:
C₈H₁₄ Boiling Pt: 131…132 °C Melting Pt: –103 °C Density: 0.7510 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009471 CAS Number: 1942-45-6 EINECS: 217-730-2 UN: 3295 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...