1,2-Dibromobenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 98%
CAS No.: 583-53-9
Molecular Formula: C6H4Br2
Formula Weight: 235.91
Flash Point: 91°(195°F)
Refractive Index: 1.6110
Odour: Strong
Melting Point: 4-6°
Boiling Point: 224-226°
Beilstein Reference: 970241
UN No.: 3082
MDL No.: MFCD00000057
Formula:
C₆H₄Br₂ Boiling Pt: 224…226 °C Melting Pt: 4…6 °C Density: 1.985 Flash Pt: 91 °C (195 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000057 CAS Number: 583-53-9 EINECS: 209-507-3 UN: 3082 ADR: 9,III |