2-Bromobenzonitrile
Supplier: Thermo Scientific
SB01215.1 SB01215.10 SB01215.50
MAYBSB01215.1EA 0 EUR
MAYBSB01215.1 MAYBSB01215.10 MAYBSB01215.50
2-Bromobenzonitrile
2-Bromobenzonitrile
Formula: C₇H₄BrN
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 51…52 °C
Density: 1.496 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001772
CAS Number: 2042-37-7
EINECS: 218-045-1
UN: 3439
ADR: 6.1,III