1-Bromo-2,4-difluorobenzene ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,4-Difluorobromobenzene
A14098.14
A14098.22
A14098.36
A14098.14EA
47.2
EUR
A14098.14
A14098.22
A14098.36
1-Bromo-2,4-difluorobenzene ≥98%
1-Bromo-2,4-difluorobenzene
Purity: 98+%
CAS No.: 348-57-2
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 51°(123°F)
Refractive Index: 1.5050
Melting Point: -4°
Boiling Point: 145-146°
Beilstein Reference: 1680892
UN No.: 1993
MDL No.: MFCD00000330
Formula:
C₆H₃BrF₂ Boiling Pt: 145…146 °C Melting Pt: –4 °C Density: 1.708 Flash Pt: 51 °C (123 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000330 CAS Number: 348-57-2 EINECS: 206-479-4 UN: 1993 ADR: 3,III |