Measuring beakers with handle
Supplier: KARTELL
1086 1087 1088 1089 1090
KART1086EA 15.7 EUR
KART1086 KART1087 KART1088 KART1089 KART1090
Measuring beakers with handle
Beakers
PP, translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 minutes), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.