Toluene-p-sulphonic anhydride 95%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 4-Methylbenzenesulphonic anhydride, p-Toluenesulphonic anhydride
367070050 367070250 367071000
ACRO367070050EA 53.7 EUR
ACRO367070050 ACRO367070250 ACRO367071000
Toluene-p-sulphonic anhydride 95%
Toluene-p-sulphonic anhydride
Formula: C₁₄H₁₄O₅S₂
MW: 326.39
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 3261
ADR: 8 III
Order Now
Specification Test Results
Appearance Off-white to beige to grey Powder or crystals and/or chunks
Infrared spectrum Conforms
Titration by coupling ≥94 %