Bromopentafluorobenzene 99%
Supplier: Thermo Fisher Scientific
Warning
107220050 107220250 107221000
ACRO107220050EA 45.9 EUR
ACRO107220050 ACRO107220250 ACRO107221000
Bromopentafluorobenzene 99%
Bromopentafluorobenzene
Formula: C₆BrF₅
Boiling Pt: 137 °C (760 mmHg)
Melting Pt: –31 °C
Density: 1.981 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00000287
CAS Number: 344-04-7
EINECS: 206-449-0
Order Now
Specification Test Results
Appearance Clear colorless to slightly colored Liquid
Infrared spectrum Conforms
Refractive index 1.4485 to 1.4505 (20°C, 589 nm)
GC ≥98.5 %
Absorption Ratio P-15/P+15 1.21 to 1.77
BSC Certification Passes