Bromopentafluorobenzene ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
A13273.14
A13273.22
A13273.36
A13273.14EA
97.8
EUR
A13273.14
A13273.22
A13273.36
Bromopentafluorobenzene ≥99%
Bromopentafluorobenzene
Purity: 99%
CAS No.: 344-04-7
Molecular Formula: C6BrF5
Formula Weight: 246.97
Flash Point: 87°(188°F)
Refractive Index: 1.4500
Melting Point: ca -31°
Boiling Point: 136-137°
Beilstein Reference: 1819390
MDL No.: MFCD00000287
Formula:
C₆BrF₅ Boiling Pt: 136…137 °C Melting Pt: ∼–31 °C Density: 1.981 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000287 CAS Number: 344-04-7 EINECS: 206-449-0 |