2-Ethylhexyl acrylate ≥98% stabilised
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Acrylic acid 2-ethylhexyl ester
L03854.AE
L03854.AP
L03854.0F
AB177784.2500
L03854.AEEA
28
EUR
L03854.AE
L03854.AP
L03854.0F
ABCRAB177784.2500
2-Ethylhexyl acrylate ≥98% stabilised
2-Ethylhexyl acrylate
Purity: 98%
CAS No.: 103-11-7
Molecular Formula: C11H20O2
Formula Weight: 184.28
Flash Point: 79°(174°F)
Refractive Index: 1.4360
Melting Point: -90°
Boiling Point: 130°/50mm
Beilstein Reference: 1765828
MDL No.: MFCD00009495
Formula:
C₁₁H₂₀O₂ Boiling Pt: 130 °C (50 mmHg) Melting Pt: –90 °C Density: 0.885 Flash Pt: 79 °C (174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009495 CAS Number: 103-11-7 EINECS: 203-080-7 |