4-(Trifluoromethyl)phenylhydrazine

Supplier: Thermo Scientific

Synonyms: 1-(4-(Trifluoromethyl)phenyl)hydrazine, 4-Hydrazinobenzotrifluoride

CD11012.1 CD11012.10
MAYBCD11012.1EA 0 EUR
MAYBCD11012.1 MAYBCD11012.10
4-(Trifluoromethyl)phenylhydrazine
4-(Trifluoromethyl)phenylhydrazine
Formula: C₇H₇F₃N₂
MDL Number: MFCD00042508
CAS Number: 368-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR