5-Bromo-2-fluorobenzaldehyde ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
A14219.09
A14219.18
A14219.30
A14219.09EA
53.7
EUR
A14219.09
A14219.18
A14219.30
5-Bromo-2-fluorobenzaldehyde ≥98%
5-Bromo-2-fluorobenzaldehyde
Purity: 98%
CAS No.: 93777-26-5
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point: 109°(228°F)
Refractive Index: 1.5700
Boiling Point: 230°
Beilstein Reference: 7632798
MDL No.: MFCD00070755
Formula:
C₇H₄BrFO Boiling Pt: 230 °C Melting Pt: 49 °C Density: 1.710 Flash Pt: 109 °C (228 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00070755 CAS Number: 93777-26-5 EINECS: 298-056-6 |