Ammonia solution 1 mol/l (1 N), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
M-Clarity™ quality level = MQ200
09859-1L 09859-250ML
SIAL09859-1LEA 152 EUR
SIAL09859-1L SIAL09859-250ML
Ammonia solution 1 mol/l (1 N), Sigma-Aldrich®
Ammonia solution
Formula: NH₃
MW: 35.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III