4-Methoxybenzonitrile, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4-Cyanoanisol, Anisonitrile, p-Cyanoanisol, p-Methoxybenzonitrile
M-Clarity™ quality level = MQ100
132470-100G
132470-25G
SIAL132470-100GEA
120
EUR
SIAL132470-100G
SIAL132470-25G
4-Methoxybenzonitrile, Sigma-Aldrich®
4-Methoxybenzonitrile
Formula:
C₈H₇NO Boiling Pt: 256 °C (1013 hPa) Melting Pt: 57…60 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001818 CAS Number: 874-90-8 EINECS: 212-871-6 UN: 3439 ADR: 6.1,III |