2-Methyl-1,3-propanediol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1,3-Dihydroxy-2-methylpropane

M-Clarity™ quality level = MQ200

375721-1L 375721-3L
SIAL375721-1LEA 43.9 EUR
SIAL375721-1L SIAL375721-3L
2-Methyl-1,3-propanediol, Sigma-Aldrich®
2-Methyl-1,3-propanediol
Formula: C₄H₁₀O₂
MDL Number: MFCD00082586
CAS Number: 2163-42-0
EINECS: 412-350-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR