Sodium tetrathionate dihydrate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Disodium tetrathionate dihydrate
M-Clarity™ quality level = MQ200
S5758-100G
S5758-25G
SIALS5758-100GEA
832
EUR
SIALS5758-100G
SIALS5758-25G
Sodium tetrathionate dihydrate, Sigma-Aldrich®
Sodium tetrathionate dihydrate
Formula:
Na₂O₆S₄·2H₂O MW: 306.27 g/mol |
MDL Number:
MFCD00150868 CAS Number: 13721-29-4 EINECS: 233-260-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...