Isobutyl cinnamate, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ300

W219304-10KG-K W219304-1KG-K
SIALW219304-10KG-KEA 1230 EUR
SIALW219304-10KG-K SIALW219304-1KG-K
Isobutyl cinnamate, Sigma-Aldrich®
Isobutyl cinnamate
Formula: C₁₃H₁₆O₂
MDL Number: MFCD00038289
CAS Number: 122-67-8
EINECS: 204-564-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR