2-Bromo-5-fluorophenol ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
A15064.06
A15064.14
AB114210.0100
A15064.06EA
41.6
EUR
A15064.06
A15064.14
ABCRAB114210.0100
2-Bromo-5-fluorophenol ≥97%
2-Bromo-5-fluorophenol
Purity: 97%
CAS No.: 147460-41-1
Molecular Formula: C6H4BrFO
Formula Weight: 191.00
Flash Point: 87°(188°F)
Refractive Index: 1.5540
Boiling Point: 184-186°
Beilstein Reference: 7915214
MDL No.: MFCD00040939
Formula:
C₆H₄BrFO Boiling Pt: 184…186 °C Density: 1.717 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040939 CAS Number: 147460-41-1 |