Jars, specimen, with ground cover glass plate, DURAN®
Supplier: DWK Life Sciences
213630504 213631106 213631303 213631903 213632805 213634703 213635802
SCOT213630504EA 55 EUR
SCOT213630504 216-0272 SCOT213631303 SCOT213631903 SCOT213632805 SCOT213634703 SCOT213635802
Jars, specimen, with ground cover glass plate, DURAN®
Jars General Use Jars
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Museum jars are used to store materials. The large glass surfaces allow a good view into the glass on its contents.