Sample containers with screw cap
Supplier: KARTELL
1672 1673 1674 1675
KART1672EA 181 EUR
KART1672 KART1673 KART1674 KART1675
Sample containers with screw cap
Sample Containers General Purpose Containers
PFA, translucent, with PFA screw cap.
  • Very good chemical resistance
  • Very good temperature resistance
For sample collection, transport and storage.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904