Benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
12010-1L-F 12010-250ML-F
SIAL12010-1L-FEA 176 EUR
SIAL12010-1L-F SIAL12010-250ML-F
Benzaldehyde, Sigma-Aldrich®
Benzaldehyde
Formula: C₆H₅CHO
MW: 106.12 g/mol
Boiling Pt: 179
Melting Pt: –26 °C
Density: 1.049 g/cm³ (20 °C)
Flash Pt: 64
Storage Temperature: Ambient
MDL Number: MFCD00003299
CAS Number: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9 III
Merck Index: 13,01057
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98811dd8cad9340872b5c4b5bfb305860eb1f81a&vid=934c1b87225e77b213e66c334d8f27ef4d924847&prid=web&clid=SAPDAM