5-Methoxysalicylaldehyde 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde
164160050 164160100 164160500
ACRO164160050EA 91.3 EUR
ACRO164160050 ACRO164160100 ACRO164160500
5-Methoxysalicylaldehyde 98%
5-Methoxysalicylaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 103 °C (2.5 mmHg)
Melting Pt: 4 °C
Density: 1.21 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0
Order Now
Specification Test Results
Appearance Clear yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20°C, 589 nm)
GC ≥97.5 %