5-Methoxysalicylaldehyde ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 6-Hydroxy-m-anisaldehyde, 2-Hydroxy-5-methoxybenzaldehyde
A15753.06 A15753.14 A15753.18 A15753.22
A15753.06EA 96.4 EUR
A15753.06 A15753.14 A15753.18 A15753.22
5-Methoxysalicylaldehyde ≥98%
5-Methoxysalicylaldehyde
Purity: 98% CAS No.: 672-13-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 110°(230°F) Refractive Index: 1.5780 Melting Point: 13-16° Boiling Point: 248-252° Beilstein Reference: 1100532 MDL No.: MFCD00003332
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 248…252 °C
Melting Pt: 13…16 °C
Density: 1.221
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003332
CAS Number: 672-13-9
EINECS: 211-589-0