VWR®, Funnels with Short Stem
Supplier: VWR Collection
221-0175EA 25.7 EUR
221-0175 221-0176 221-0177 221-0178 221-0179 221-0180 221-0181 221-0182
VWR®, Funnels with Short Stem
Funnels Filtration Funnels Filter Funnels
Borosilicate 3.3 glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Smooth internal walls.
Certifications: ISO 4798.